Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD70
Supplier:  Biolegend 355107
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD70 [113-16]; Isotype: Mouse IgG1; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7411