Learn More
Biolegend PerCP/Cyanine5.5 anti-human Ganglioside GD2
Supplier:  Biolegend 357311
Encompass_Preferred
PerCP/Cyanine5.5 anti-human Ganglioside GD2 [14G2a]; Isotype: Mouse IgG2a, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-207-7658