Learn More
Biolegend PerCP/Cyanine5.5 anti-human HLA-DR
Supplier:  Biolegend 327019
Encompass_Preferred
PerCP/Cyanine5.5 anti-human HLA-DR [LN3]; Isotype: Mouse IgG2b, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: FC; Size: 25 tests
Catalog No. 50-207-5743