Learn More
Biolegend PerCP/Cyanine5.5 anti-human IL-10
Supplier:  Biolegend 501418
Encompass_Preferred
PerCP/Cyanine5.5 anti-human IL-10 [JES3-9D7]; Isotype: Rat IgG1, κ; Reactivity: Human, Cross-Reactivity: Baboon, Rhesus, Cynomolgus; Apps: ICFC; Size: 100 tests
Catalog No. 50-208-5524