Learn More
Biolegend PerCP/Cyanine5.5 anti-human TCR Vγ9
Supplier:  Biolegend 331322
Encompass_Preferred
PerCP/Cyanine5.5 anti-human TCR Vγ9 [B3]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. 50-207-6012