Learn More
Biolegend PerCP/Cyanine5.5 anti-human Arginase I [14D2C43]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Supplier:  Biolegend 369709
Encompass_Preferred
PerCP/Cyanine5.5 anti-human Arginase I [14D2C43]; Isotype: Mouse IgG2b, κ; Reactivity: Human; Apps: ICFC; Size: 25 tests
Catalog No. 50-237-6528