Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD32B/C
Supplier:  Biolegend 398310
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD32B/C [S18005H]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-207-9365