Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD25
Supplier:  Biolegend 985804
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD25 [M-A251]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 500 uL
Catalog No. 50-213-2098