Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD103 (Integrin αE)
Supplier:  Biolegend 350226
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD103 (Integrin αE) [Ber-ACT8]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus; Apps: FC; Size: 100 tests
Catalog No. NC1744400