Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD79b (Igβ)
Supplier:  Biolegend 132809
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD79b (Igβ) [HM79-12]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0963