Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse IL
Catalog No. 502721890
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 μg
Catalog No. 50-272-1890 Supplier Biolegend Supplier No. 505027
PerCP/Cyanine5.5 anti-mouse IL-10 [JES5-16E3]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: ICFC; Size: 25 μg