Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse NK-1.1
Supplier:  Biolegend 156526
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse NK-1.1 [S17016D]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-213-1820