Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD117 (c-kit)
Supplier:  Biolegend 135134
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD117 (c-kit) [ACK2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1094