Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD152
Supplier:  Biolegend 106315
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD152 [UC10-4B9]; Isotype: Armenian Hamster IgG; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-206-9942