Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD244.2 (2B4 B6 Alloantigen)
Supplier:  Biolegend 133513
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD244.2 (2B4 B6 Alloantigen) [m2B4 (B6)458.1]; Isotype: Mouse IgG1, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-0969