Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD49d
Supplier:  Biolegend 103619
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD49d [R1-2]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-206-9778