Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD63
Supplier:  Biolegend 143912
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD63 [NVG-2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, FC; Size: 100 ug
Catalog No. 50-207-1484