Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD88 (C5aR)
Supplier:  Biolegend 135812
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD88 (C5aR) [20/70]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 ug
Catalog No. 50-207-1148