Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse/human CD207 (Langerin) [4C7]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Human; Apps: FC; Size: 25 ug
Supplier:  Biolegend 144215
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse/human CD207 (Langerin) [4C7]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse, Human; Apps: FC; Size: 25 ug
Catalog No. 50-237-6900