Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse CD276 (B7-H3)
Supplier:  Biolegend 135616
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse CD276 (B7-H3) [MIH35]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-1144