Learn More
Biolegend PerCP/Cyanine5.5 anti-mouse IgD
Supplier:  Biolegend 405710
Encompass_Preferred
PerCP/Cyanine5.5 anti-mouse IgD [11-26c.2a]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. 50-207-9744