Learn More
Biolegend PerCP/Cyanine5.5 anti-rat IgM [MRM-47]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC; Size: 100 ug
Supplier:  Biolegend 408916
Encompass_Preferred
PerCP/Cyanine5.5 anti-rat IgM [MRM-47]; Isotype: Mouse IgG; Reactivity: Rat; Apps: FC; Size: 100 ug
Catalog No. 50-207-9858