Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD33
Supplier:  Biolegend 366615
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD33 [P67.6]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. NC1834801