Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD61
Supplier:  Biolegend 336418
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD61 [VI-PL2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. NC1931253