Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Human CD14 (61D3) ..... Cat. No. 65-0149-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 650149T025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Human CD14 (61D3) ..... Cat. No. 65-0149-T025 ..... Size 25 testsThe 61D3 antibody is specific for human CD14, a 53-55 kDa GPI-anchored glycoprotein. CD14 is highly expressed on monocytes and to a lesser extent on interfollicular macrophages, and some dendritic cells. Together with LPS-Binding Protein (LBP), CD14 binds to and mediates the innate immune response to bacterial lipopolysaccharide (LPS).
Catalog No. 50-210-3570