Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Human CD274 PD-L1 (B7-H1) (29E.2A3) ..... Cat. No. 65-5984-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 655984T025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Human CD274 PD-L1 (B7-H1) (29E.2A3) ..... Cat. No. 65-5984-T025 ..... Size 25 testsThe 29E.2A3 antibody reacts with human CD274 which is also known as PD-L1 or B7-H1. CD274 is a type 1 transmembrane glycoprotein and a member of the Ig superfamily. PD-L1 is a ligand for CD279 (PD-1) and plays an important role in regulating immune cell responses. CD274 is expressed on activated monocytes, macrophages, dendritic cells, and T cells.
Catalog No. 50-210-3605