Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Human CD200 (OX-104) ..... Cat. No. 65-9200-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 659200T025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Human CD200 (OX-104) ..... Cat. No. 65-9200-T025 ..... Size 25 testsThe OX-104 monoclonal antibody reacts with human CD200, a 40-45kD transmembrane glycoprotein that is also known as OX2. CD200 is a member of the Ig superfamily and is expressed by various cell types, including B cells, a subset of T cells, follicular dendritic cells, endothelial cells, neurons, and a subset of CD34 positive progenitor cells. The interaction between CD200 and its receptor, CD200R, is of importance in the control of macrophage and granulocyte activation and may contribute to pathways that suppress and limit their activity in a variety of tissues, suggesting a regulatory function for CD200.
Catalog No. 50-210-3606