Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Human CD5 (UCHT2) ..... Cat. No. 65-0059-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 650059T025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Human CD5 (UCHT2) ..... Cat. No. 65-0059-T025 ..... Size 25 testsThe UCHT2 antibody is specific for human CD5, a 67 kD transmembrane glycoprotein, that is expressed on most thymocytes, mature T cells, and a subset of B cells. CD5 is involved in modulating antigen receptor signaling in both T and B cells and binds to CD72, expressed on B cells.
Catalog No. 50-210-3564