Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Human CD56 (NCAM) (MY31) ….. Cat. No. 65-0564-T100 ….. Size 100 tests
Supplier:  Cytek Biosciences Inc 650564T100
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Human CD56 (NCAM) (MY31) ….. Cat. No. 65-0564-T100 ….. Size 100 testsThe MY31 antibody reacts with human CD56, also known as the neural cell adhesion molecule (NCAM), a glycoprotein which is a member of the immunoglobulin superfamily. The 140 kDa isoform of CD56 is expressed on human NK cells and NK-T cells, with increased expression levels on activated NK lymphocytes. The CD56 antigen is also expressed by neurons and is reported to play a role in nervous system development and neural cell-to-cell adhesion.Clone MY31 also reacts with a subset of CD14+ monocytes in non-human primates, and is reported to be cross-reactive with Chimpanzee, Cynomolgus and Rhesus.
Catalog No. 50-210-3587