Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Human CD69 (FN50) ..... Cat. No. 65-0699-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 650699T025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Human CD69 (FN50) ..... Cat. No. 65-0699-T025 ..... Size 25 testsThe FN50 monoclonal antibody reacts with human CD69, a type II transmembrane glycoprotein also known as the Very Early Activation Antigen, EA-1, Leu23, Activation Inducer Molecule (AIM) and CLEC2C. CD69 is expressed as a disulfide-linked homodimer on activated T and B cells, NK cells, neutrophils, and monocytes. Induction occurs rapidly upon activation and is transient. It is also constitutively expressed on platelets and a subset of thymocytes.
Catalog No. 50-210-3591