Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Mouse CD134 (OX40) (OX-86) ..... Cat. No. 65-1341-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 651341U025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Mouse CD134 (OX40) (OX-86) ..... Cat. No. 65-1341-U025 ..... Size 25 µgThe OX-86 monoclonal antibody reacts with mouse CD134, a 50kD type 1 integral membrane glycoprotein and a member of the TNF-receptor superfamily. CD134 is also known as OX40 and is expressed on activated CD4 and CD8 T cells. CD134 is the receptor for CD252 (OX40 ligand) and the interaction of this receptor-ligand pair is implicated in T-dependent humoral responses as well as numerous aspects of T cell expansion, survival and memory. Additionally, CD134 seems to play a significant role in the pathogenesis of some autoimmune diseases. 
Catalog No. 50-210-3597