Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Mouse CD252 (OX40 Ligand) (RM134L) ..... Cat. No. 65-5905-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 655905U025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Mouse CD252 (OX40 Ligand) (RM134L) ..... Cat. No. 65-5905-U025 ..... Size 25 µgThe RM134L antibody recognizes CD252, also known as OX-40 Ligand or CD134 Ligand, a member of the TNF-superfamily that is present on the surface of antigen presenting cells and activated B lymphocytes. The OX-40 Ligand interacts with OX-40 (CD134) which is expressed primarily on activated T cells. This costimulatory interaction leads to increased proliferation and IL-2 production responses of activated T cells, and at the same time enhances proliferation and immunoglobulin secretion by activated B cells.The RM134L antibody is useful for flow cytometric detection of CD252 on stimulated mouse splenocytes. It has also been reported to block the costimulatory activity of OX-40 Ligand. Please choose the appropriate format for each application.
Catalog No. 50-210-3600