Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Mouse CD309 (FLK1) (Avas12a1) ..... Cat. No. 65-5821-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 655821U025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Mouse CD309 (FLK1) (Avas12a1) ..... Cat. No. 65-5821-U025 ..... Size 25 µgThe Avas12a1 monoclonal antibody reacts with mouse Flk-1, also known as vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a receptor tyrosine kinase involved in vascular endothelial tissue development and is expressed on endothelial cells during embryonic stages and some endothelial tissues in the adult.
Catalog No. 50-210-3599