Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Mouse CD70 (FR70) ..... Cat. No. 65-0701-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 650701U025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Mouse CD70 (FR70) ..... Cat. No. 65-0701-U025 ..... Size 25 µgThe FR70 monoclonal antibody reacts with mouse CD70, a 30-33 kDa type II transmembrane glycoprotein. CD70 is a member of the TNF-family and is expressed by activated mouse T and B lymphocytes. Interaction of CD70 with CD27, its ligand on T cells, is important in T-B cell crosstalk and plays a role in costimulation and immune regulation.
Catalog No. 50-210-3592