Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Mouse CD90.2 (30-H12) ..... Cat. No. 65-0903-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 650903U025
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Mouse CD90.2 (30-H12) ..... Cat. No. 65-0903-U025 ..... Size 25 µgThe 30-H12 antibody reacts with mouse CD90.2 (Thy1.2). CD90.2 is a strain-specific allelic form of the GPI-linked membrane associated protein CD90 and is involved in adhesion and signal transduction. CD90.2 is expressed on thymocytes, mature T cells and neurons in mouse strains that express the CD90.2 allele (BALB/c, CBA, C3H, C57BL/6, SJL and others). 30-H12 does not react with the CD90.1 allele expressed in mouse strains such as PL and AKR.
Catalog No. 50-210-3593