Learn More
Cytek Biosciences Inc Antibodies PerCP-Cyanine5.5 Anti-Mouse Ly-6C (HK1.4) ….. Cat. No. 65-5932-U100 ….. Size 100 µg
Supplier:  Cytek Biosciences Inc 655932U100
Encompass_Preferred
Antibodies PerCP-Cyanine5.5 Anti-Mouse Ly-6C (HK1.4) ….. Cat. No. 65-5932-U100 ….. Size 100 µgThe HK1.4 monoclonal antibody reacts with mouse Ly-6C, a 15-17kD GPI-linked cell-surface protein. Ly-6C is expressed on monocyte/macrophage cells, endothelial cells, granulocytes and a subset of lymphocytes. Mice strains with the Ly-6.2 alloantigen have subsets of both Ly-6C positive CD4 and CD8 cells while Ly-6.1 expressing strains have only the CD8 Ly-6C positive subset.
Catalog No. 50-210-3603