Learn More
Biolegend PerCP/Cyanine5.5 anti-Tubulin β 3 (TUBB3)
Supplier:  Biolegend 801216
Encompass_Preferred
PerCP/Cyanine5.5 anti-Tubulin β 3 (TUBB3) [TUJ1]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Mouse, Rat; Apps: ICFC; Size: 100 tests
Catalog No. 50-208-9844