Learn More
Biolegend PerCP/FireTM 780 anti-human
Supplier:  Biolegend 342937
Encompass_Preferred
PerCP/Fire™ 780 anti-human TCR Vα24-Jα18 (iNKT cell) [6B11]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: FC; Size: 25 tests
Catalog No. 50-253-7917