Learn More
Biolegend PerCP/FireTM 780 anti-mouse CD
Supplier:  Biolegend 161224
Encompass_Preferred
PerCP/Fire™ 780 anti-mouse CD62L [W18021D]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-279-6578