Learn More
Biolegend PerCP/FireTM 780 anti-mouse CD
Catalog No. 502906296
Encompass_Preferred
PerCP/Fire™ 780 anti-mouse CD31 (PECAM-1) [W18222B]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-290-6296 Supplier Biolegend Supplier No. 160217
PerCP/Fire™ 780 anti-mouse CD31 (PECAM-1) [W18222B]; Isotype: Rat IgG2b, κ; Reactivity: Mouse; Apps: FC; Size: 25 μg