Learn More
Biolegend PerCP/FireTM 780 anti-mouse/hu
Catalog No. 502906257
Encompass_Preferred
PerCP/Fire™ 780 anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human; Apps: FC; Size: 100 μg
Catalog No. 50-290-6257 Supplier Biolegend Supplier No. 103214
PerCP/Fire™ 780 anti-mouse/human CD45R/B220 [RA3-6B2]; Isotype: Rat IgG2a, κ; Reactivity: Mouse, Human; Apps: FC; Size: 100 μg