Learn More
Biolegend PerCP/FireTM 806 anti-human
Supplier:  Biolegend 359150
Encompass_Preferred
PerCP/Fire™ 806 anti-human CD195 (CCR5) [J418F1]; Isotype: Rat IgG2b, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. 50-253-7889