Learn More
Biolegend PerCP/FireTM 806 anti-human CD
Catalog No. 502722331
Encompass_Preferred
PerCP/Fire™ 806 anti-human CD39 [A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-272-2331 Supplier Biolegend Supplier No. 328250
PerCP/Fire™ 806 anti-human CD39 [A1]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests