Learn More
Biolegend PerCP/FireTM 806 anti-mouse CD
Catalog No. 502796486
Encompass_Preferred
PerCP/Fire™ 806 anti-mouse CD127 (IL-7Rα) [S18006K]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-279-6486 Supplier Biolegend Supplier No. 158221
PerCP/Fire™ 806 anti-mouse CD127 (IL-7Rα) [S18006K]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 25 μg