Learn More
Biolegend PerCP/FireTM 806 anti-mouse CD
Supplier:  Biolegend 158222
Encompass_Preferred
PerCP/Fire™ 806 anti-mouse CD127 (IL-7Rα) [S18006K]; Isotype: Rat IgG2b, λ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-279-6535