Learn More
ABclonal Technology PerCP Rabbit anti-Mouse CD45
SDP
Catalog No. 5000010898
Encompass_Preferred
This gene encodes a protein with heparan sulfate proteoglycan binding heparin binding and protein tyrosine phosphatase activities It plays roles in lymphocyte differentiation regulation of macromolecule metabolism and signal transduction It is involved in processes such as lymphocyte activation and protein phosphorylation The protein is located on the plasma membrane focal adhesions and membrane rafts and is expressed in structures like the 3rd branchial arch alimentary system cardiovascular system and placenta It is used in studies of systemic lupus erythematosus Its human ortholog is linked to diseases like hepatitis C multiple sclerosis and severe combined immunodeficiency It is orthologous to human PTPRC (protein tyrosine phosphatase receptor type C)
Catalog No. 50-000-10898 Supplier ABclonal Technology Supplier No. A27305200T
This gene encodes a protein with heparan sulfate proteoglycan binding heparin binding and protein tyrosine phosphatase activities It plays roles in lymphocyte differentiation regulation of macromolecule metabolism and signal transduction It is involved in processes such as lymphocyte activation and protein phosphorylation The protein is located on the plasma membrane focal adhesions and membrane rafts and is expressed in structures like the 3rd branchial arch alimentary system cardiovascular system and placenta It is used in studies of systemic lupus erythematosus Its human ortholog is linked to diseases like hepatitis C multiple sclerosis and severe combined immunodeficiency It is orthologous to human PTPRC (protein tyrosine phosphatase receptor type C)