Learn More
Biolegend PerCP/Cyanine5.5 anti-human CD57
Supplier:  Biolegend 359622
Encompass_Preferred
PerCP/Cyanine5.5 anti-human CD57 [HNK-1]; Isotype: Mouse IgM, κ; Reactivity: Human; Apps: FC; Size: 100 tests
Catalog No. NC1098010