Learn More
Abcam PerCP/Cy5.5 Conjugation Kit - Lightning-Link, 100UG
Catalog No. 502585584
Encompass_Preferred
Labeling in < 4 hrs with 30 secs hands-on time using PerCP/Cy5.5 Conjugation Kit - Lightning-Link ab102911. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5584 Supplier Abcam Supplier No. AB102911100UG
Labeling in < 4 hrs with 30 secs hands-on time using PerCP/Cy5.5 Conjugation Kit - Lightning-Link ab102911. 100% antibody recovery.
The product is subject to the following: Abcam Restricted Use Statement