Learn More
Cytek Biosciences Inc PERCPCY5.5 M CD3E(145-2C11)25U
Supplier:  Cytek Biosciences Inc 650031U025
Encompass_Preferred
PerCP-Cyanine5.5 Anti-Mouse CD3e (145-2C11) Isotype: Armenian Hamster IgG Apps: FC Size: 25 ug
Catalog No. 50-105-4910