Learn More
Biolegend PerCP/Cyanine5.5 anti-rat CD90/mouse CD90.1 (Thy-1.1)
Supplier:  Biolegend 202516
Encompass_Preferred
PerCP/Cyanine5.5 anti-rat CD90/mouse CD90.1 (Thy-1.1) [OX-7]; Isotype: Mouse IgG1, κ; Reactivity: Rat, Mouse (AKR/J and PL mouse strains).; Apps: FC; Size: 100 ug
Catalog No. 50-165-390